Orders LinkContactUsLinkComponent
Masterflex® Sanitary Tees
Masterflex® Sanitary Tees
Nome del fornitore:   Avantor Fluid Handling
Masterflex® Sanitary Tees
Nome del fornitore:  Avantor Fluid Handling
Certificates
Masterflex® Sanitary Tees
6 varianti
Product Details & Documents
These compliant, biocompatible fittings provide assurance, integrity, and cost-effective solutions.
  • Engineered to meet the needs of bioprocess workflows, single-use tees uphold the highest standards of compliance to meet critical needs
  • Created for flawless sterility, sanitary tees can undergo gamma irradiation and autoclaving, ensuring decontamination in all processes
  • Constructed from USP Class VI materials, sanitary tees prioritise biocompatibility and safety, guaranteeing their suitability for sensitive bioprocess applications
  • Designed for easy integration, single-use tees are engineered to provide reliable connections and consistent performance
Sanitary single-use tees are engineered and manufactured from gamma irradiatable and autoclavable Polypropylene and PVDF USP Class VI materials. These fittings offer an uncompromising commitment to sterilisation. Manufactured within an ISO Class 7 cleanroom, these sanitary tees provide the highest level of biocompatibility and safety, ensuring seamless integration into critical processes within the biopharmaceutical industry.
Specifications for Product Family
Specifiche
  • Sterilizzazione:
  • Gamma, Autoclave
  • Gamma, Autoclave
  • Gamma, Autoclave
  • Gamma, Autoclave
  • Gamma, Autoclave
  • Gamma, Autoclave
Specifiche
Product Family Options
Informazioni sul prodotto
  • Tipo di connessione
    Fitting material
    Sanitary clamp size (Port 1)
    Disponibilità
    Price
  • MFLX40618-76
    Fitting, Sanitary Tee, ¾", Polypropylene; EA
    Fitting, Sanitary Tee, ¾", Polypropylene; EA
    Sanitary
    Polypropylene
    ¾"
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 95.30
    SFr. 95.30
    1 * 1 ST
       SFr. 95.30
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX40618-76
    Sterilizzazione
    Gamma, Autoclave
  • MFLX40618-78
    Fitting, Sanitary Tee, 1", Polypropylene; EA
    Fitting, Sanitary Tee, 1", Polypropylene; EA
    Sanitary
    Polypropylene
    1"
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 107.00
    SFr. 107.00
    1 * 1 ST
       SFr. 107.00
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX40618-78
    Sterilizzazione
    Gamma, Autoclave
  • MFLX40618-80
    Fitting, Sanitary Tee, 1½", Polypropylene; EA
    Fitting, Sanitary Tee, 1½", Polypropylene; EA
    Sanitary
    Polypropylene
    1½"
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 113.00
    SFr. 113.00
    1 * 1 ST
       SFr. 113.00
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX40618-80
    Sterilizzazione
    Gamma, Autoclave
  • MFLX40618-82
    Fitting, Sanitary Tee, ¾", PVDF; EA
    Fitting, Sanitary Tee, ¾", PVDF; EA
    Sanitary
    PVDF
    ¾"
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 138.00
    SFr. 138.00
    1 * 1 ST
       SFr. 138.00
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX40618-82
    Sterilizzazione
    Gamma, Autoclave
  • MFLX40618-84
    Fitting, Sanitary Tee, 1", PVDF; EA
    Fitting, Sanitary Tee, 1", PVDF; EA
    Sanitary
    PVDF
    1"
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 171.00
    SFr. 171.00
    1 * 1 ST
       SFr. 171.00
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX40618-84
    Sterilizzazione
    Gamma, Autoclave
  • MFLX40618-86
    Fitting, Sanitary Tee, 1½", PVDF; EA
    Fitting, Sanitary Tee, 1½", PVDF; EA
    Sanitary
    PVDF
    1½"
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 224.00
    SFr. 224.00
    1 * 1 ST
       SFr. 224.00
    Specifiche:
    More
    Specifications
    Cat. No.
    MFLX40618-86
    Sterilizzazione
    Gamma, Autoclave