Cerex® FLORASIL SPE Columns
Column FLORASIL, 6 ml, 1000 mg
Codice catalogo TCAT30213512
Nome del fornitore: Tecan
Vai a
Product Details & Documents
Florisil is used for sample preparation (solid phase extraction) of e.g., organic tin compounds and aliphatic carboxylic acids. Florisil is a magnesium silicate.
- Ion-exchange
- Liquid handler compatible
Specifiche
- Capacità:6 ml
- Descrizione:Column FLORASIL
- Peso letto:1000 mg