Anti-KIR2DS2 Rabbit Polyclonal Antibody
Codice catalogo PRSI8201
Nome del fornitore: ProSci Inc.
Specifiche
- Simbolo dell'antigene:KIR2DS2
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Rabbit
- ImmunoChimica:Yes
- ImmunoFluorescenza:Yes
- Isotipo:IgG
- Reattività:Human
- Western blot:Yes
- Forma:Liquid
- ID gene:100132285
- Sinomini antigene:p58 natural killer cell receptor clone CL-49|p58 NK receptor|Killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 2|NKAT5|Natural killer-associated transcript 5|p58 KIR|MHC class I NK cell receptor|KIR2DL3
- Tampone di conservazione:Supplied in PBS containing 0,02% sodium azide.
- Peso molecolare:34, 42 kDa
- Temperatura di conservazione:KIR2DS2 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
- Concentrazione:1 mg/ml
- Immunogeno:KIR2DS2 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human KIR2DS2. The immunogen is located within the last 50 amino acids of KIR2DS2.
- Applicazioni testate:ELISA, immunocytochemistry, immunofluorescence, western blot.
- Purificazione:KIR2DS2 antibody is affinity chromatography purified via peptide column.
- Taglia:0,02/0,1 mg
- Conf:100 µG
Specifiche
Informazioni su questo articolo
Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells (1). The KIR proteins are classified by the number of extracellular immunoglobulin domains and by whether they have a long (L) or short (S) cytoplasmic domain (2,3). KIR proteins are thought to play an important role in regulation of the immune response (3). KIR2DS2 downregulates the cytotoxicity of NK cells upon recognition of specific class I major histocompatibility complex (MHC) molecules on target cells and is a receptor on natural killer (NK) cells for HLA-C alleles (3,4).
Type:
Antigen: KIR2DS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human