Orders LinkContactUsLinkComponent
Maxitips, Pipette Tips | Avantor
Maxitips, Pipette Tips
   Nachhaltige Optionen verfügbar
Anbieter:   EPPENDORF
Maxitips, Pipette Tips
Anbieter:  EPPENDORF
   Nachhaltige Optionen verfügbar
Zertifikate
Maxitips, Pipette Tips
3 Varianten
Product Details & Documents
Eppendorf Maxitips are designed for large volume liquid handling, offering high precision and compatibility with Eppendorf pipetting systems. Ideal for diverse lab applications, they ensure accuracy and ease of use, enhancing workflow efficiency.
  • Use Maxitips P to aspirate 1 to 10 ml from beakers using the positive displacement principle
  • Use Maxitips S to aspirate between 2,5 and 10 ml from high, narrow-mouth vessels with the air cushion principle
  • Valve for Maxitips ensures drip-free dispensing of liquids with a high vapour pressure
  • Graduation on Maxitips ensures accurate dispensing of non-aqueous liquids
Experience exceptional levels of flexibility with different Maxipettor pipette tips that allow liquid removal from a variety of vessels. When aspirating from small vessels such as beakers, select the Eppendorf Maxitip P for precise aspiration of 1 to 10ml volumes. For tall, narrow-necked vessels and measuring flasks, use the longer-tip Maxitip S to aspirate volumes of 2,5 to 10 ml.
Designed for use with Maxipettor large volume pipette models from Eppendorf, the Maxitips pipette tips are perfect for all large vessel types.
Bestellinformationen: Please note: Maxitips are designed for use with Eppendorf Maxipettor® 4720 only.
Spezifikationen für diese Produktfamilie
Technische Daten
  • Catalog No:
  • 612-7321
  • 612-7333
  • 612-7331
  • Environmentally Preferable:
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Produzione a basso impatto
  • Certificazione ambientale:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Technische Daten
Product Family Options
Produktinformationen
  • Stile punta
    Volume
    Confezione
    Verfügbarkeit
    Preis
  • Maxitips P
    1 - 10 ml
    100 pieces
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 100 ST
    1 * 100 ST - SFr. 145.00
    SFr. 145.00
    1 * 100 ST
       SFr. 145.00
    Spezifikationen:
      Produzione a basso impatto
    Mehr
    Specifications
    Cat. No.
    612-7321
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • Maxitips S
    2,5 - 10 ml
    200 pieces
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 200 ST
    1 * 200 ST - SFr. 104.00
    SFr. 104.00
    1 * 200 ST
       SFr. 104.00
    Spezifikationen:
      Produzione a basso impatto
    Mehr
    Specifications
    Cat. No.
    612-7333
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto
  • Maxitips S starter kit
    2,5 - 10 ml
    100 Maxitips, 10 dispensing Parts, 10 valves
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 KIT
    1 * 1 KIT - SFr. 110.00
    SFr. 110.00
    1 * 1 KIT
       SFr. 110.00
    Spezifikationen:
      Produzione a basso impatto
    Mehr
    Specifications
    Cat. No.
    612-7331
    Environmentally Preferable
    Produzione a basso impatto
    Certificazione ambientale
    Renewable Energy Certificate, ISO 14001
      Produzione a basso impatto