Snooded Polypropylene Caps
Snooded PP caps, bianco
Codice catalogo HOPE70.401
Nome del fornitore: Medicom
Vai a
Product Details & Documents
These snooded caps provide extra protection for workers with long hair in industrial environments.
- Fully protects and contains long hair
- Conveys professional image
- Ability to use color code for tracking
- Single use
Informazioni per l'ordine: Includes caps and dispenser box.
Documents
TDS PP Snooded Peaked Cap.pdf
Scheda dati
Specifiche
- Colore:bianco
- Descrizione:Snooded PP caps
- Cat. No.:HOPE70.401
- Peso:40 g/m²
- Applicazioni per abiti:Laboratory|Industrial