Human Tagged ORF Clone, SPCA2 (ATP2C2)
Catalog # ORIGRG239585
Supplier: OriGene
CAS Number:
Specifications
- Conf:10 µG
- Descrizione:SPCA2 (ATP2C2) (NM_001291454) human tagged ORF clone
- Vector details:pCMV6-AC-GFP
Specifications
About this item
ATP2C2 (tGFP-tagged) - Human ATPase, Ca++ transporting, type 2C, member 2 (ATP2C2), transcript variant 3.
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence. This clone is substantially in agreement with the reference, but a complete review of all prevailing variants is recommended prior to use.