Orders LinkContactUsLinkComponent
Bottles, HDPE, with Thread | Avantor
Bottles, HDPE, with Thread
Bottles, HDPE, with Thread
Zertifikate
Bottles, HDPE, with Thread
2 Varianten
Product Details & Documents
Round bottles made up of robust HDPE plastic, black.
  • With retracted labeling field
  • Tasteless and odorless
  • Ideal for filling chemical-technical substances
Zertifizierung: UN approval for liquids. For high-quality filling goods that require UN approval , ideal for retention patterns.
Product Family Options
Produktinformationen
  • Capacità
    Descrizione
    Filettatura
    Colore
    Altezza
    Verfügbarkeit
    Preis
  • 500 ml
    Bottles, round, 50 g weight
    RD 45
    nero
    176,5 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 10 ST
    1 * 10 ST - SFr. 60.90
    SFr. 60.90
    1 * 10 ST
       SFr. 60.90
  • 1000 ml (1 L)
    Bottles, round, 88 g weight
    RD 45
    nero
    211,8 mm
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 10 ST
    1 * 10 ST - SFr. 88.40
    SFr. 88.40
    1 * 10 ST
       SFr. 88.40