Communicating tubes
765-0969
- Descrizione:Communicating tube with foot
- Conf:1
Five glass tubes of different shape and inner dimensions. Down to each other. When filling the liquid in the pipes, it is clear that the height of the stowage in the tubes. pipes are independent of the design of the pipes. Comes with foot.