6-Azaindole
Glass bottle for solids, 1 g
Codice catalogo MOLE66418762-1G
Marketsource
Nome del fornitore: Molekula
6-Azaindole
Codice catalogo MOLE66418762-1G
Nome del fornitore: Molekula
Marketsource
Numero CAS: 271-29-4
Vai a
Product Details & Documents
- Numero CAS:271-29-4
- Formula:C₇H₆N₂
- Numero MDL:MFCD01686899
- Molecular Weight:118.14 g/mol
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:1 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



