Orders LinkContactUsLinkComponent
Chenodeoxycholic acid 97+%
Chenodeoxycholic acid 97+%
Nome del fornitore:   Thermo Fisher Scientific
Chenodeoxycholic acid 97+%
Nome del fornitore:  Thermo Fisher Scientific
Certificates
Chenodeoxycholic acid 97+%
3 varianti
Product Details & Documents
Sinonimi:  3α,7α-Dihydroxycholanic acid, 7α-Hydroxylithocholic acid, 7alpha-Hydroxylithocholic acid, 3α,7α-Dihydroxy-5β-cholan-24-oic acid, 3α,7α-Dihydroxycholan-24-oic acid, Chenodiol, 3α,7α-Dihydroxy-5β-cholanic acid
  • Numero CAS:474-25-9
  • Formula:C₂₄H₄₀O₄
  • Numero MDL:MFCD00064142
  • Molecular Weight:392.58 g/mol
  • IndexMerck:12,02096
  • Punto di fusione:162…168 °C
  • Densità:1,162 g/cm³ (20 °C)
  •  :Density: 1.162 g/cm³ (20 °C)
  • temperatura di conservazione:Ambiente
  • EINECS:207-481-8
Documents
Specifications for Product Family
Risultati dei test sulle specifiche
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤2 %
  • Specific optical rotation:
    +9° to +14° (20°C, 589 nm) (c=2, EtOH)
  • Titration with NaOH:
    ≥97.0 % (on dried basis)