Skin, block model
Skin structure
Catalog # 763-0123
Marketsource
Supplier: 3B Scientific
Jump to:
Product Details & Documents
70× enlargement in the model of skin with hair from the head, skin with hair from the armpit and skin from a finger. Sweat glands, sensory bodies, etc. are inlaid in the three sections. Dimensions: 30×36×32 cm.
Specifications
- Descrizione:Skin structure