Storage Cabinets for Non-flammable Media
Catalog # DUEH20-200967-050
Supplier: DUPERTHAL
Specifications
- Descrizione:Storage cabinets for non-flammable media
- L×P×A:894×612×2045 mm
- Porte:Wing door
- Conf:1
Specifications
About this item
The storage of solid and liquid, non-flammable and non-corrosive chemicals and is specifically marked for the storage of toxic substances.
- Sheet steel cabinet with hinged doors, painted in RAL 7004 (signal gray)
- Equipment: Hinged doors with twist handle security lock, closing on 3 sides for safe access
- Floor collecting tray made of galvanized sheet steel according to StawaR
- 3x perforated storage floor made of galvanized sheet steel
- Height adjustable with anti-lift device, Adjustable feet for level adjustment
Certifications: Type testing by TÜV SÜD according to furniture standards DIN EN 16121 and DIN EN 16122.