- Descrizione:Winlog.Validation evaluation software for ebro data loggers
- Conf:1
- Per:Pharmaceutical and medical application
The easy-to-use software products are suitable for programming and read-out of Ebro® data loggers as well as for the evaluation of the measured values. For routine checks and validation.
- User-friendly
- Highly accurate measurements
- Automatic report generation
- Automatic user-defined calculations
- Automatic recognition of process cycles
- Current version: 4.0
Save time and effort for the regularly required process validation with the help of the powerful reporting and evaluation software Winlog.validation. The automatic recognition of the individual process steps makes your work easier and meets the high demands of validation and qualification in pharmacy and medicine.
High-performance report and evaluation software for the stringent requirements of validation and qualification in the pharmaceutical and medical sectors. Supports: EBI 12, EBI 10, EBI 11, EBI 16, EBI 20, EBI 40, EBI 100, EBI 300, EBI 310.
Creation of user-defined templates for specific devices and thermal processes. Three-dimensional display of sensor placement. FDA 21. CFR Part 11 functionality also possible. IQ-/OQ documentation available. Automatic completion of validation processes.
Hardware requirements: Processor speed minimum 1,6 GHz, working memory 2 GB (32 bit), working memory 4 GB (64 bit) 4 GB free hard disc space, USB A (universal serial bus).
Software requirements: Windows 8 (32 bit or 64 bit), windows 10 (32 bit or 64 bit), windows 11.