Orders LinkContactUsLinkComponent
Digital pH Sensor, Memosens CPS71E
Digital pH Sensor, Memosens CPS71E
Nome del fornitore:   Endress+Hauser
Digital pH Sensor, Memosens CPS71E
Nome del fornitore:  Endress+Hauser
Certificates
Digital pH Sensor, Memosens CPS71E
Digital pH Sensor, Memosens CPS71E
3 varianti
Product Details & Documents
With its contamination-resistant reference system, Memosens CPS71E is a digital pH sensor for highly contaminated or toxic media such as strong acids or bases.
  • The special ion trap of the electrode prevents poisoning of the junction and the reference, offers resistance to strong acids and bases and ensures a longer service life of the sensor
  • The optional pressurized reference ensures reliable measurements in blocking media
  • Optional upside-down mounting for flexible installation
  • Inductive cable connection and non-contact signal transmission improve process integrity by preventing problems with moisture or corrosion
  • On-site sensor replacement minimizes process downtime and costs
Memosens 2.0 technology enables trend identification, predictive maintenance and advanced IIoT services with extended data storage for calibration and process information.
Informations de commande: For optional items or more information, contact your Avantor representative.
Informations de livraison: Digital pH sensor Memosens CPS71E.
Product Family Options
Non hazardous area electrodes
Informazioni sul prodotto
  • pH
    Temperatura
    Lunghezza
    Disponibilità
    Price
  • ENDRCPS71E-1009/0
    Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), non-hazardous area approval
    Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), non-hazardous area approval
    0 - 14 pH
    0...140 °C
    120 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - 612.00 SFr.
    612.00 SFr.
    1 * 1 ST
       612.00 SFr.
  • ENDRCPS71E-1265
    Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), non-hazardous area approval
    Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), non-hazardous area approval
    0 - 12 pH
    0...140 °C
    120 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - 630.00 SFr.
    630.00 SFr.
    1 * 1 ST
       630.00 SFr.
ATEX electrodes
Informazioni sul prodotto
  • pH
    Temperatura
    Lunghezza
    Disponibilità
    Price
  • ENDRCPS71E-BA7BTB2
    Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), ATEX II 1G Ex ia IIC T3/T4/T6 Ga
    Digital pH sensor Memosens CPS71E, 0,8 - 14 bar (abs), ATEX II 1G Ex ia IIC T3/T4/T6 Ga
    0 - 14 pH
    0...140 °C
    120 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - 630.00 SFr.
    630.00 SFr.
    1 * 1 ST
       630.00 SFr.