Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Carboxyrhodamine 6G (5-CR6G)
5-Carboxyrhodamine 6G (5-CR6G) single isomer ≥95%
5-Carboxyrhodamine 6G (5-CR6G) single isomer ≥95%
5-Carboxyrhodamine 6G (5-CR6G) single isomer ≥95%
Catalog # AATB331
Supplier:
AAT Bioquest
CAS Number:
Expand image
5-Carboxyrhodamine 6G (5-CR6G) single isomer ≥95%
Catalog # AATB331
Supplier:
AAT Bioquest
CAS Number:
Specifications
Conf:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
CAS Number:
180144-69-8
Product Details