ContactUsLinkComponent
TRITC-Dextran Conjugate (Average MW = ~70K)
TRITC-Dextran Conjugate (Average MW = ~70K)
Codice catalogo AATB21708
Nome del fornitore:  AAT Bioquest
Numero CAS:  
undefined
TRITC-Dextran Conjugate (Average MW = ~70K)
Codice catalogo AATB21708
Nome del fornitore:  AAT Bioquest
Numero CAS:  
Specifiche
  • Conf:
    25 mg
  • Descrizione:
    TRITC–dextran conjugate (average MW = ~70K)
Specifiche
Informazioni su questo articolo
TRITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.
  • High quality
  • Multiple applications
Caution: For research use only.