Amplite® Red Peroxidase Substrate
Catalog # AATB11011
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:1.000 Assays
- Descrizione:Amplite™ Red
Specifications
About this item
Amplite® Red is a sensitive fluorogenic peroxidase substrate that generates a highly red fluorescent product that has maximum absorption of 571 nm and maximum emission of 585 nm.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.