ReadiLink™ Rapid mFluor™ Violet 540 Antibody Labeling Kit "Microscale Optimized for Labeling 50 µg Antibody Per Reaction" (2 Labelings)
Catalog # AATB1114
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Conf:2 Tests
- Descrizione:ReadiLink™ rapid mFluor™ violet 540 antibody labelling kit 'Microscale optimized for labelling 50 µg antibody per reaction'
Specifications
About this item
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
- Violet-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.