Amplite® Colorimetric Glucose Oxidase Assay Kit
AATB11299
:
- Conf:500 Tests
- Descrizione:Amplite™ colorimetric glucose oxidase assay kit
The glucose oxidase is a dimeric protein that catalyses the oxidation of beta-D-glucose into hydrogen peroxide and D-glucono-1,5-lactone, which is hydrolysed to gluconic acid.
- In-house QC testing
- Sensitive
- Suggested protocol available
: For research use only.