Amplite® Colorimetric Glucose-6-Phosphate Dehydrogenase (G6PD) Assay Kit
Catalog # AATB13807
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:200 Tests
- Descrizione:Amplite™ colorimetric glucose-6-phosphate dehydrogenase (G6PD) assay kit
Specifications
About this item
Glucose-6-phosphate dehydrogenase (G6PD) catalyses the conversion of glucose-6-phosphate to 6-phosphoglucono-δ-lactone, the first and rate-limiting step in the pentose phosphate pathway.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.