StrandBrite™ Green Fluorimetric RNA Quantitation Kit *Optimised for Microplate Readers*
Codice catalogo AATB17655
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:1.000 Tests
- Descrizione:StrandBrite™ green fluorimetric RNA quantitation kit *optimized for microplate readers*
Specifiche
Informazioni su questo articolo
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, reverse transcription PCR, and differential display PCR.
- Highly sensitive RNA detection
- Robust protocol
- Multiple applications
Caution: For research use only.