Cell Navigator® Lysosome Staining Kit *Deep Red Fluorescence*
Codice catalogo AATB22659
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:500 Tests
- Descrizione:Cell Navigator™ lysosome staining kit *deep red fluorescence*
Specifiche
Informazioni su questo articolo
Lysosomes are cellular organelles which contain acid hydrolase enzymes to break up waste materials and cellular debris.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Attenzione: For research use only.