ReadiLink™ iFluor® 647 FISH Fluorescence Imaging Kit
Katalognr. AATB17316
Anbieter: AAT BIOQUEST
CAS-Nummer:
Technische Daten
- Conf:25 Tests
- Descrizione:Convenient tool for labelling a target DNA using an iFluor® 647 labeled FISH probe via in situ hybridization, for PCR microplates
Technische Daten
Über diesen Artikel
The kit provides Taq DNA polymerase enzyme which incorporates iFluor® 647-dUTPs in the target DNA through Polymerase Chain Reaction (PCR).
- Robust protocol
- Bright labeling
- Photostable fluorescence
The Readilink™ iFluor® 647 FISH fluorescence imaging kit is a convenient tool for labelling a target DNA using an iFluor® 647 labeled FISH probe via in situ hybridisation..
Our proprietary iFluor® dyes are brighter and more photostable than traditional fluorescent labels, providing the desired resolution and signal.
Packaging: Packaged in plastic vials.
Caution: For research use only.