PhosphoWorks™ Colorimetric MESG Phosphate Assay Kit *UV Absorption*
Katalognr. AATB21659
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:200 Tests
- Descrizione:PhosphoWorks™ colorimetric MESG phosphate assay kit *UV absorption*
Specifications
About this item
In the presence of inorganic phosphate MESG is converted to 2-amino-6-mercapto-7-methlpurine by purine nucleoside phosphorylase (EC 2.4.2.1) with absorption wavelength shift to red.
- Provides all necessary components
- Multiple applications
- Photometric detection
Caution: For research use only.