Cell Meter™ Live Cell TUNEL Apoptosis Assay Kit *Red Fluorescence* (50 Tests)
Codice catalogo AATB22844
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:50 Tests
- Descrizione:Cell Meter™ live cell TUNEL apoptosis assay kit *red fluorescence*
Specifiche
Informazioni su questo articolo
DNA fragmentation represents a characteristic of late stage apoptosis.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Attenzione: For research use only.