ReadiLink™ Rapid mFluor™ Violet 510 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Catalog # AATB1110
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Conf:2 Tests
- Descrizione:ReadiLink™ rapid mFluor™ violet 510 antibody labeling kit microscale optimised for labeling 50 µg antibody per reaction
Specifications
About this item
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
- Violet-laser excitable
- Robust and simple antibody labelling protocol
- No column purification required
Caution: For research use only.