ReadiUse™ Preactivated HRP Maleimide
Codice catalogo AATB11026
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:300 µG
- Descrizione:ReadiUse™ preactivated HRP maleimide
Specifiche
Informazioni su questo articolo
ReadiUse™ Preactivated maleimide is a thiol-reactive HRP derivative.
- Ready-to-use
- Highly useful antibody label
- Widely used for immunoassays
- Multiple applications
Caution: For research use only.