Signal Guard™ HRP Conjugate Stabilizer
Catalog # AATB11010
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Conf:50 mL
- Descrizione:Signal Guard™ HRP conjugate stabilizer
Specifications
About this item
The major advantage of a liquid HRP-conjugate is that it eliminates the chance for human error when dissolving or diluting the lyophilized or concentrated conjugate.
- Permits liquid-hrp conjugates to be stored at 4°c for 24 months or RT for 6 months
- Can create stock solutions or ready-to-use reagents
Caution: For research use only.