Amplite® Fluorimetric Proteasome 20S Activity Assay Kit *Green Fluorescence*
Catalog # AATB13456
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:100 Tests
- Descrizione:Amplite™ Fluorimetric proteasome 20S activity assay kit *green fluorescence*
- Test per kit:100 Tests
Specifications
About this item
Our Amplite® Fluorometric 20S Proteasome Assay Kit is a homogeneous fluorescent assay that measures the chymotrypsin-like protease activity associated with the proteasome complex either in cultured cells or cell lysates.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.