Amplite® Colorimetric L-Lactate Dehydrogenase (LDH) Assay Kit
Catalog # AATB13813
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:200 Tests
- Descrizione:Amplite™ Colorimetric L-lactate dehydrogenase (ldh) assay kit
- Test per kit:200 Tests
Specifications
About this item
Lactate dehydrogenase (LDH) is an oxidoreductase enzyme that catalyses the interconversion of pyruvate and lactate.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.