Amplite® Fluorimetric Glucose Oxidase Assay Kit *Red Fluorescence*
Catalog # AATB11300
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:500 Tests
- Descrizione:Amplite™ fluorimetric glucose oxidase assay kit *red fluorescence*
- Test per kit:500 Tests
Specifications
About this item
The glucose oxidase is a dimeric protein that catalyses the oxidation of beta-D-glucose into hydrogen peroxide and D-glucono-1,5-lactone, which is hydrolyzed to gluconic acid.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.