Amplite® Colorimetric D-Lactate Assay Kit
Catalog # AATB13811
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:200 Tests
- Descrizione:Amplite™ colorimetric d-lactate assay kit
- Test per kit:200 Tests
Specifications
About this item
Lactic acid is chiral and has two optical isomers: L-lactic acid and D-lactic acid.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.