Amplite® Colorimetric Hydrogen Peroxide Assay Kit
Catalog # AATB11500
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:500 Tests
- Descrizione:Amplite™ colorimetric hydrogen peroxide assay kit
- Test per kit:500 Tests
Specifications
About this item
Hydrogen peroxide (H2O2) is a reactive oxygen metabolic by-product that serves as a key regulator for a number of oxidative stress-related states.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.