Amplite® Fluorimetric Glucose-6-Phosphate Dehydrogenase (G6PD) Assay Kit
Codice catalogo AATB13806
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:200 Tests
- Descrizione:Amplite™ fluorimetric glucose-6-phosphate dehydrogenase (G6PD) assay kit
- Test per kit:200 Tests
Specifiche
Informazioni su questo articolo
Glucose-6-phosphate dehydrogenase (G6PD) catalyses the conversion of glucose-6-phosphate to 6-phosphoglucono-δ-lactone, the first and rate-limiting step in the pentose phosphate pathway.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.