Amplite® Colorimetric Malondialdehyde (MDA) Quantitation Kit
Codice catalogo AATB10070
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:200 Tests
- Descrizione:Amplite™ colorimetric malondialdehyde (MDA) quantitation kit
- Test per kit:200 Tests
Specifiche
Informazioni su questo articolo
Malondialdehyde (MDA) is natural byproduct of lipid peroxidation and is widely used as a key indicator to determine the oxidative stress and free radical formation.
- In-house QC testing
- Sensitive
- Suggested protocol available
Caution: For research use only.