Auftragserfassung
Switzerland
ContactUsLinkComponent
Cell Meter™ Cell Proliferation Assay Kit
  AATB22505
 :  
undefined
Cell Meter™ Cell Proliferation Assay Kit
  AATB22505
 :  
  • Conf:
    200 Tests
  • Test per kit:
    200 Tests
The kit does not require cell lysis, long incubations or removal of cells with minimal hands-on time. It is compatible with high throughput screening.
  • bright fluorescence
  • green color
  • improved accuracy
The kit uses our Nuclear Green LCS1, a proprietary fluorescent dye that exhibits strong fluorescence upon binding to nucleic acids. The use of Nuclear Green LCS1 improves accuracy over cell metabolism-based cell proliferation or cytotoxicity assays that can be influenced by cell changes that are unrelated to cell numbers.
Sensitive fluorescence-based method for quantifying cells and assessing cell proliferation and cytotoxicity in a microplate format