Cell Navigator® Lysosome Staining Kit *Green Fluorescence*
Catalog # AATB22656
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:500 Tests
- Descrizione:Cell Navigator™ Lysosome staining kit *green fluorescence*
- Test per kit:500 Tests
Specifications
About this item
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labeling sub-cellular organelles such as membranes, lysosomes, mitochondria, nuclei and etc.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Caution: For research use only.