Screen Quest™ Fluo-8 Medium Removal Calcium Assay Kit, Optimized for Difficult Cell Lines
Nome del fornitore: AAT Bioquest
Certificati
Informazioni su questo articolo
Calcium flux assays are preferred methods in drug discovery for screening G protein coupled receptors (GPCR).
- 100 to 250 times fluorescence increase once bound to calcium
- Contains all necessary components
- Suitable for multiple types of cells
- Robust and adaptable protocol
- Brighter and more photostable signal
Caution: For research use only.
Informazioni correlate
Manual Fluo-8 Medium Removal Calcium Assay Kit, Optimized for Difficult Cell Line.pdf Manual Screen Quest Fluo 8 Medium Removal Calcium Assay Kit Optimized for Difficult Cell Lines.pdf Manual Screen Quest™ Fluo 8 Medium Removal Calcium Assay Kit Optimized For Difficult Cell Lines.pdf Manual Screen Quest™ Fluo-8 Medium Removal Calcium Assay Kit Optimized for Difficult Cell Lines.pdf
Manuale
Frequently Bought Together