Spexyte™ Micro pH Probe *Optimized for Nanodrop*
Codice catalogo AATB21230
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:500 Tests
- Descrizione:Spexyte™ Micro pH probe *Optimized for Nanodrop*
- Test per kit:500 Tests
Specifiche
Informazioni su questo articolo
Biological samples are usually in a very narrow pH range from 4.5 to 8.5, and many of them need to be provided in micro-volume (microliter).
- Suitable for micro-volume samples
- Sensitive and quick results
- Fits four-parameter logistic model
Caution: For research use only.