Cell Navigator® Cell Plasma Membrane Staining Kit *Green Fluorescence*
AATB22682
:
- Conf:500 Tests
- Descrizione:Cell Navigator™ cell plasma membrane staining kit *green fluorescence*
- Test per kit:500 Tests
The Cell Navigator® Plasma Membrane Stain Kit provides a fast and uniform labeling of the plasma membrane without the cell-type differences exhibited by lectins.
- Signal persists through fixation
- Stable labeling regardless of cell type
- Multiple applications
: For research use only.