Cell Navigator® Live Cell Endoplasmic Reticulum (ER) Staining Kit *Blue Fluorescence*
Catalog # AATB22634
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:100 Tests
- Descrizione:Cell Navigator™ live cell endoplasmic reticulum (ER) staining kit *blue fluorescence*
- Test per kit:100 Tests
Specifications
About this item
The endoplasmic reticulum (ER) is a type of organelle in the cells of eukaryotic organisms that forms an interconnected network of flattened, membrane-enclosed sacs or tube-like structures known as cisternae.
- Cell-permeant staining
- Improved signal
- Suitable for all major fluorescence platforms
Caution: For research use only.