Live or Dead™ Fixable Dead Cell Staining Kit *Red Fluorescence*
Codice catalogo AATB22603
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:200 Tests
- Descrizione:Live or Dead™ fixable dead cell staining kit *red fluorescence*
- Test per kit:200 Tests
Specifiche
Informazioni su questo articolo
Our Live or Dead™ Fixable Dead Cell Staining Kits are a set of tools for labeling cells for fluorescence microscopic investigations of cell functions.
- Contains all necessary components
- Bright and stable signal
- Suitable for all major fluorescence platforms
Attenzione: For research use only.