Cell Meter™ Live Cell Caspase 9 Binding Assay Kit *Green Fluorescence*
Codice catalogo AATB20117
Nome del fornitore: AAT Bioquest
Numero CAS:
Specifiche
- Conf:25 Tests
- Descrizione:Cell Meter™ Live cell Caspase 9 binding assay kit *Green fluorescence*
- Test per kit:25 Tests
Specifiche
Informazioni su questo articolo
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
- Based on FMK inhibitor
- Direct caspase detection
- Suitable for all major fluorescence platforms
Attenzione: For research use only.