Cell Meter™ Mitochondrial Hydroxyl Radical Detection Kit *Red Fluorescence*
Catalog # AATB16055
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:200 Tests
- Descrizione:Cell Meter™ Mitochondrial hydroxyl radical detection kit *red fluorescence*
- Test per kit:200 Tests
Specifications
About this item
The detection of intracellular hydroxyl radical is of central importance to understanding proper cellular redox regulation and the impact of its dysregulation on various pathologies.
- High sensitivity
- Bright fluorescence
- Suitable for live cell imaging
- Detection in 1 hour
Caution: For research use only.