Cell Meter™ Live Cell Caspase 3/7 Binding Assay Kit *Green Fluorescence*
Catalog # AATB20100
Supplier: AAT Bioquest
CAS Number:
Specifications
- Conf:25 Tests
- Descrizione:Cell Meter™ live cell caspase 3/7 binding assay kit *Green fluorescence*
- Test per kit:25 Tests
Specifications
About this item
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
- Based on FMK inhibitor
- Direct caspase detection
- Suitable for all major fluorescence platforms
Caution: For research use only.