Cell Meter™ Intracellular Fluorimetric Hydrogen Peroxide Assay Kit *Green Fluorescence*
Catalog # AATB11503
Anbieter: AAT Bioquest
CAS-Nummer:
Technische Daten
- Conf:200 Tests
- Descrizione:Cell Meter™ intracellular fluorimetric hydrogen peroxide assay kit *Green Fluorescence*
- Test per kit:200 Tests
Technische Daten
Über diesen Artikel
Hydrogen peroxide (H₂O₂) is a reactive oxygen metabolic by-product that serves as a key regulator for a number of oxidative stress-related states.
- Mix-and-read format
- Adaptable to hts instrumentation
- Quantifies h2o2 in live cells
Caution: For research use only.