Orders LinkContactUsLinkComponent
Cell Meter™ Fixable Viability Dye, RX660
Cell Meter™ Fixable Viability Dye, RX660
Cell Meter™ fixable viability dye, 200 Tests, 200 Tests
Codice catalogo AATB22530
Nome del fornitore:   AAT Bioquest
Cell Meter™ Fixable Viability Dye, RX660
Codice catalogo AATB22530
Nome del fornitore:  AAT Bioquest
Numero CAS:  
Cell Meter™ Fixable Viability Dye, RX660
Cell Meter™ Fixable Viability Dye, RX660
Cell Meter™ fixable viability dye, 200 Tests, 200 Tests
Numero CAS:  
Price
300.00 SFr.
Log in per disponibilità
1 * 200 Tests
Product Details & Documents
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-Impermeable
  • Amine- And Thiol-Reactive
  • High Stability
  • Live/Dead Cell Labeling Dye
  • Fixable
Caution: For research use only.
Specifiche
Specifiche
  • Conf:
    200 Tests
  • Descrizione:
    Cell Meter™ fixable viability dye
  • Test per kit:
    200 Tests