Order Entry
Switzerland
ContactUsLinkComponent
 
Irritant
APC-Cy5.5 Tandem fluorescent probe
Catalog # AATB2622
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
APC-Cy5.5 Tandem fluorescent probe
Catalog # AATB2622
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Tipo di conf.:
    Plastic bottle
  • Conf:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :105000 g/mol
ADR:5,II
 :GHS07