Orders LinkContactUsLinkComponent
Normal grinding, laboratory kit, NS29
Normal grinding, laboratory kit, NS29
Distillation Set, Macro 22
Codice catalogo 201-0210
Nome del fornitore:   witeg Labortechnik
Normal grinding, laboratory kit, NS29
Codice catalogo 201-0210
Nome del fornitore:  witeg Labortechnik
Normal grinding, laboratory kit, NS29
Normal grinding, laboratory kit, NS29
Distillation Set, Macro 22
Price
1 260.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Supplementary kit for 171021. Consists of the following components:
1 pc Round flask 1000 ml, NS 29/32
1 round flask 1000 ml, 3 necks
2 pcs Capillary 6 mm
1 pc Thermometer 0 + 250 ° C
1 Dest publisher Anschütz-Thiele NS 29/32 - NS 19/26
1 pc Dest column Hempel 300 mm, NS 29/32
1 st Dest column Column Vigreaux 400 mm, NS 29/32
1 evacuation essay NS 29/32
1 Gas Wash Bottle 250 ml, complete
Specifiche
  • Descrizione:
    Distillation Set, Macro 22