Orders LinkContactUsLinkComponent
Cleanroom Socks
Cleanroom Socks
Supplier:   ISOFIELD IRELAND
Cleanroom Socks
Supplier:  ISOFIELD IRELAND
Certificates
Cleanroom Socks
3 Options Available Below
Product Details & Documents
Non-sterile, lightweight, single-use cleanroom socks.
  • 95% polyester, 5% spandex
  • Mid-calf length
  • White colour
  • Three sizes for added flexibility
  • Low linting
Store in a dry, cool place (<40 °C) away from direct sunlight. Do not expose open cartons to prolonged direct fluorescent light.
Cleanroom socks are designed for single-use application in pharmaceutical product preparation, cleanroom cleaning and preparation, medical device manufacturing, blending operations for biotechnologies, handling of active materials, and maintenance of equipment and instruments.
Shelf life: Five years from date of manufacture.
Certifications: ISO Class 4 and 5 cleanroom, Class 10 and 100 cleanroom. Manufactured in a facility operating under ISO 9001:2015 quality management system. Processed in a NEBB certified ISO Class 5 cleanroom.
Packaging: 10 pairs per double bag, 50 sealed outer cleanroom bags per carton, 500 pairs per carton.
Specifications for Product Family
Specifications
  • Catalog No:
  • ISOF70610S-M
  • ISOF70610XXL
  • ISOF70610L-XL
  • Classe di cleanroom:
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • Applicazioni per abiti:
  • Pharmaceutical|Laboratory
  • Pharmaceutical|Laboratory
  • Pharmaceutical|Laboratory
Specifications
Product Family Options
Product Information
  • Colore
    Taglia dell'abbigliamento
    Availability
    Price
  • bianco
    S/M
    Log in for availability
    Log in for availability
    1 * 500 PAIRE
    1 * 500 PAIRE - SFr. 622.00
    SFr. 622.00
    1 * 500 PAIRE
       SFr. 622.00
    Specifications:
    More
    Specifications
    Cat. No.
    ISOF70610S-M
    Classe di cleanroom
    10/ISO 4
    Applicazioni per abiti
    Pharmaceutical|Laboratory
  • bianco
    M/L
    Log in for availability
    Log in for availability
    1 * 500 PAIRE
    1 * 500 PAIRE - SFr. 915.00
    SFr. 915.00
    1 * 500 PAIRE
       SFr. 915.00
    Specifications:
    More
    Specifications
    Cat. No.
    ISOF70610XXL
    Classe di cleanroom
    10/ISO 4
    Applicazioni per abiti
    Pharmaceutical|Laboratory
  • bianco
    L/XL
    Log in for availability
    Log in for availability
    1 * 500 PAIRE
    1 * 500 PAIRE - SFr. 622.00
    SFr. 622.00
    1 * 500 PAIRE
       SFr. 622.00
    Specifications:
    More
    Specifications
    Cat. No.
    ISOF70610L-XL
    Classe di cleanroom
    10/ISO 4
    Applicazioni per abiti
    Pharmaceutical|Laboratory